Novus Biologicals
Manufacturer Code:NBP270625AF647
Catalog # N270625A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C4 |
Immunogen | Recombinant protein fragment corresponding to amino acids 496-723 of human EHHADH (NP_001957) produced in E.coli. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 3,2-trans-enoyl-CoA isomerase; 3-hydroxyacyl-CoA dehydrogenase; Enoyl-CoA hydratase/3,2-trans-enoyl-CoA isomerase; enoyl-CoA hydratase/3-hydroxyacyl CoA dehydrogenase enoyl-Coenzyme A hydratase/3-hydroxyacyl Coenzyme A dehydrogenase LBFP LBP L-PBE PBFEMGC120586 peroxisomal peroxisomal bifunctional enzyme32-trans-enoyl-CoA isomerase peroxisomal enoyl-CoA hydratase; enoyl-Coenzyme A, hydratase/3-hydroxyacyl Coenzyme A dehydrogenase; L-3-hydroxyacyl-CoA dehydrogenase; L-bifunctional protein; L-bifunctional protein, peroxisomal; LBP; MFE1; Multifunctional enzyme 1; PBE; Peroxisomal bifunctional enzyme; peroxisomal enoyl-CoA hydratase
Gene Aliases: ECHD; EHHADH; FRTS3; L-PBE; LBFP; LBP; PBFE
UniProt ID: (Human) Q08426
Entrez Gene ID: (Human) 1962
Molecular Function:
dehydrogenase
epimerase/racemase
hydratase
isomerase
lyase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.