Invitrogen
Manufacturer Code:PA570659
Catalog # PIPA570659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of rat EHD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EH domain-containing protein 3; EH-domain containing 3; EH-domain containing protein 2; PAST homolog 3; PAST3 EH domain containing 3 EH domain-containing protein 3 PAST homolog 3
Gene Aliases: EHD2; EHD3; PAST3; si:ch211-147c1.2
UniProt ID: (Human) Q9NZN3, (Rat) Q8R491, (Mouse) Q9QXY6
Entrez Gene ID: (Horse) 100070691, (Bovine) 510183, (Rabbit) 100344135, (Dog) 611260, (Human) 30845, (Rat) 192249, (Mouse) 57440, (Guinea pig) 100731115, (Zebrafish) 562949
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.