Novus Biologicals
Manufacturer Code:NBP287339
Catalog # NB034961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human EHD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EH domain containing 3 EH domain-containing protein 3 EHD2 EH-domain containing 3 PAST homolog 3 PAST3; EH domain-containing protein 3; EH-domain containing 3; PAST homolog 3
Gene Aliases: EHD2; EHD3; PAST3
UniProt ID: (Human) Q9NZN3
Entrez Gene ID: (Human) 30845
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.