Invitrogen
Manufacturer Code:PA5116554
Catalog # PIPA5116554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 244-543 of human EHD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC027084 C130052H20Rik EH domain containing 2 EH domain-containing protein 2 Ehd2 EH-domain containing 2 FLJ96617 MGEPS PAST homolog 2 PAST2 putative eps protein; EH domain-containing protein 2; EH-domain containing 2; PAST homolog 2; putative eps protein
Gene Aliases: BC027084; C130052H20Rik; EHD2; MGEPS; PAST2
UniProt ID: (Human) Q9NZN4, (Rat) Q4V8H8, (Mouse) Q8BH64
Entrez Gene ID: (Human) 30846, (Rat) 361512, (Mouse) 259300
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.