Novus Biologicals
Manufacturer Code:NBP298789JF646
Catalog # NB055513
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human EHD2. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EH domain containing 2 EH domain-containing protein 2 EH-domain containing 2 FLJ96617 PAST homolog 2 PAST2; EH domain-containing protein 2; EH-domain containing 2; PAST homolog 2
Gene Aliases: EHD2; PAST2
UniProt ID: (Human) Q9NZN4
Entrez Gene ID: (Human) 30846
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.