Abnova
Manufacturer Code:H00030846A01
Catalog # 89-106-825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 365 a.a. to 427 a.a) partial recombinant protein with GST tag.,EHD2 (NP_055416 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant EHD2.
For Research Use Only
Protein Aliases: EH domain-containing protein 2; EH-domain containing 2; FLJ96617; PAST homolog 2; PAST2
Gene Aliases: EHD2; PAST2
UniProt ID: (Human) Q9NZN4
Entrez Gene ID: (Human) 30846
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.