Invitrogen
Manufacturer Code:PA566865
Catalog # PIPA566865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human EHD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EH domain-containing protein 1; EH domain-containing protein 1 PAST homolog 1 hPAST1 Testilin EHD1 PAST PAST1; EH-domain containing 1; hPAST1; PAST homolog 1; Testilin
Gene Aliases: CDABP0131; EHD1; H-PAST; HPAST1; PAST; PAST1
UniProt ID: (Human) Q9H4M9
Entrez Gene ID: (Human) 10938
Molecular Function: G-protein modulator calcium-binding protein enzyme modulator membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.