Proteintech
Manufacturer Code:246571AP150UL
Catalog # 50-173-4475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EHD1 Fusion Protein Ag18400 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EH domain containing 1 EH domain containing protein 1 EHD1 H PAST HPAST1 PAST PAST homolog 1 PAST1 Testilin; EH domain-containing protein 1; EH-domain containing 1; hPAST1; mPAST1; PAST homolog 1; Testilin
Gene Aliases: AA409636; CDABP0131; EHD1; H-PAST; HPAST1; PAST; PAST1; RGD1306960; RME-1
UniProt ID: (Human) Q9H4M9, (Mouse) Q9WVK4, (Rat) Q641Z6
Entrez Gene ID: (Human) 10938, (Mouse) 13660, (Rat) 293692
Molecular Function:
G-protein modulator
calcium-binding protein
enzyme modulator
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.