Novus Biologicals
Manufacturer Code:NBP270624V
Catalog # NBP270624V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B12 |
Immunogen | Recombinant protein fragment corresponding to amino acids 156-476 of human EGR2 (NP_000390) produced in E.coli. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AT591; AT591 CMT1D CMT4E early growth response 2 early growth response 2 (Krox-20 homolog Drosophila) early growth response protein 2 EGR-2 FLJ14547 Krox-20 homolog Drosophila KROX-20 Drosophila homolog (early growth response-2) KROX20DKFZp686J1957 Zinc finger protein Krox-20; E3 SUMO-protein ligase EGR2; E3 SUMO-protein transferase ERG2; Early growth response protein 2; EGR-2; KROX-20, Drosophila, homolog (early growth response-2); Zinc finger protein Krox-20
Gene Aliases: AT591; CMT1D; CMT4E; EGR2; KROX20
UniProt ID: (Human) P11161
Entrez Gene ID: (Human) 1959
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.