Invitrogen
Manufacturer Code:PA527814
Catalog # PIPA527814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 260 and 451 of Human EGR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AT591; AT591 CMT1D CMT4E KROX20 E3 SUMO-protein ligase EGR2 EGR-2 KROX-20 Drosophila homolog (early growth response-2) Krox-20 homolog Drosophila early growth response protein 2 zinc finger protein Krox-20; E3 SUMO-protein ligase EGR2; E3 SUMO-protein transferase ERG2; Early growth response protein 2; EGR-2; KROX-20, Drosophila, homolog (early growth response-2); Zinc finger protein Krox-20
Gene Aliases: AT591; CMT1D; CMT4E; Egr-2; EGR2; Krox-20; KROX20; NGF1-B; Zfp-25; Zfp-6
UniProt ID: (Human) P11161, (Mouse) P08152
Entrez Gene ID: (Human) 1959, (Mouse) 13654
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.