Invitrogen
Manufacturer Code:PA538588
Catalog # PIPA538588
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human EGR1/2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AT225; AT591; E3 SUMO-protein ligase EGR2; E3 SUMO-protein transferase ERG2; Early growth response protein 1; Early growth response protein 2; EGR-1; EGR-2; KROX-20, Drosophila, homolog (early growth response-2); Krox-24 nuclear protein; Nerve growth factor-induced protein A; NGFI-A; TIS8 AT225 G0S30 NGFI-A ZNF225 KROX-24 ZIF-268 EGR1 EGR2 early growth response 2 (Krox-20 homolog Drosophila) CMT1D CMT4E DKFZp686J1957 FLJ14547 KROX20 KROX-20 Drosophila homolog early growth response-2 Krox-20 homolog; Transcription factor ETR103; Transcription factor Zif268; Zinc finger protein 225; Zinc finger protein Krox-20; Zinc finger protein Krox-24
Gene Aliases: A530045N19Rik; AT225; AT591; CMT1D; CMT4E; egr; Egr-1; Egr-2; EGR1; EGR2; ETR103; G0S30; Krox-1; Krox-20; KROX-24; KROX20; KROX24; Ngf1; NGF1-A; NGF1-B; Ngfi; NGFI-A; NGFIA; TIS8; Zenk; Zfp-25; Zfp-6; ZIF-268; Zif268; ZNF225
UniProt ID: (Human) P18146, (Human) P11161, (Rat) P08154, (Mouse) P08046, (Mouse) P08152, (Rat) P51774
Entrez Gene ID: (Human) 1958, (Human) 1959, (Rat) 24330, (Mouse) 13653, (Mouse) 13654, (Rat) 114090
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.