Abnova
Manufacturer Code:H00001958M04
Catalog # 89-021-403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G7 |
Immunogen | EGR1 (NP_001955 444 a.a. ~ 543 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant EGR1.
For Research Use Only
Protein Aliases: AT225; Early growth response protein 1; EGR-1; G0S30; KROX-24; Nerve growth factor-induced protein A; NGFI-A; TIS8; Transcription factor ETR103; Transcription factor Zif268; ZIF-268; Zinc finger protein 225; Zinc finger protein Krox-24; ZNF225
Gene Aliases: AT225; EGR1; G0S30; KROX-24; KROX24; NGFI-A; TIS8; ZIF-268; ZNF225
UniProt ID: (Human) P18146
Entrez Gene ID: (Human) 1958
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.