Novus Biologicals
Manufacturer Code:NBP130440
Catalog # NBP130440
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | EG188e/d5 |
Immunogen | Partial recombinant human PHD3/HIF Prolyl Hydroxylase 3 (UniProt# Q9H6Z9) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 1.14.11 EC 1.14.11.- EGL nine (C.elegans) homolog 3 egl nine homolog 3 egl nine homolog 3 (C. elegans) egl nine-like protein 3 isoform FLJ21620 HIF prolyl hydroxylase 3 HIF-PH3 HIFPH3 MGC125998 HIF-prolyl hydroxylase 3 HPH-1 HPH-3 Hypoxia-inducible factor prolyl hydroxylase 3 MGC125999 pdh3 PHD3 Prolyl hydroxylase domain-containing protein 3; Egl nine homolog 3; egl nine-like protein 3 isoform; egl-9 family hypoxia-inducible factor 3; HIF prolyl hydroxylase 3; HIF-PH3; HIF-prolyl hydroxylase 3; HPH-1; HPH-3; Hypoxia-inducible factor prolyl hydroxylase 3; PHD3; Prolyl hydroxylase domain-containing protein 3; Prolyl hydroxylase EGLN3
Gene Aliases: EGLN3; HIFP4H3; HIFPH3; PHD3
UniProt ID: (Human) Q9H6Z9
Entrez Gene ID: (Human) 112399
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.