R&D Systems
Manufacturer Code:MAB7680100
Catalog # MAB7680100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2445B |
Immunogen | <em>S. frugiperda</em> insect ovarian cell line <em>Sf 21</em>-derived recombinant human EGLN1/PHD2 Ala2-Phe426 Accession No. Q9GZT9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rabbit Monoclonal Antibody
Protein Aliases: C1orf12 C1orf12HPH2 EC 1.14.11 EC 1.14.11.- ECYT3 egl nine homolog 1 egl nine homolog 1 (C. elegans) egl nine-like protein 1 HIF prolyl hydroxylase 2 HIFPH2 HIF-PH2 HIFPH2 EGL nine (C.elegans) homolog 1 HIF-prolyl hydroxylase 2 HPH2 HPH-2 Hypoxia-inducible factor prolyl hydroxylase 2 PHD2 PHD2SM20 Prolyl hydroxylase domain-containing protein 2 SM20 SM-20 zinc finger MYND domain-containing protein 6 ZMYND6 ZMYND6DKFZp761F179; Egl nine homolog 1; egl nine-like protein 1; egl-9 family hypoxia-inducible factor 1; HIF prolyl hydroxylase 2; HIF-PH2; HIF-prolyl hydroxylase 2; Hypoxia-inducible factor prolyl hydroxylase 2; PHD2; Prolyl hydroxylase domain-containing protein 2; SM-20; zinc finger MYND domain-containing protein 6
Gene Aliases: C1orf12; ECYT3; EGLN1; HALAH; HIF-PH2; HIFPH2; HPH-2; HPH2; PHD2; PNAS-118; PNAS-137; SM20; ZMYND6
UniProt ID: (Human) Q9GZT9
Entrez Gene ID: (Human) 54583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.