Novus Biologicals
Manufacturer Code:NB100138AF647
Catalog # NB100138Y
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to a C-terminal portion of human PHD2/HIF Prolyl Hydroxylase 2 (between amino acids 350-426) (GeneID 54583). |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf12HPH2 EC 1.14.11 EC 1.14.11.- ECYT3 egl nine homolog 1 egl nine homolog 1 (C. elegans) egl nine-like protein 1 HIF prolyl hydroxylase 2 HIF-PH2 HIFPH2 EGL nine (C.elegans) homolog 1 HIF-prolyl hydroxylase 2 HPH-2 Hypoxia-inducible factor prolyl hydroxylase 2 PHD2SM20 Prolyl hydroxylase domain-containing protein 2 SM-20 zinc finger MYND domain-containing protein 6 ZMYND6DKFZp761F179; Egl nine homolog 1; egl nine-like protein 1; egl-9 family hypoxia-inducible factor 1; HIF prolyl hydroxylase 2; HIF-PH2; HIF-prolyl hydroxylase 2; Hypoxia-inducible factor prolyl hydroxylase 2; PHD2; Prolyl hydroxylase domain-containing protein 2; SM-20; zinc finger MYND domain-containing protein 6
Gene Aliases: C1orf12; ECYT3; EGLN1; HALAH; HIF-PH2; HIFPH2; HPH-2; HPH2; PHD2; PNAS-118; PNAS-137; SM20; ZMYND6
UniProt ID: (Human) Q9GZT9
Entrez Gene ID: (Human) 54583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.