Invitrogen
Manufacturer Code:MA513697
Catalog # PIMA513697
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Type | Antibody |
Clone | 528 199.12 |
Immunogen | Partially purified EGFR from A431 cells (clone 528) and extracellular domain of recombit human EGFR protein (clone 199.12) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Antibody Cocktail
For Research Use Only
Protein Aliases: avian erythroblastic leukemia viral (v-erb-b) oncogene homolog; cell growth inhibiting protein 40; cell proliferation-inducing protein 61; Epidermal growth factor receptor; erb-b2 receptor tyrosine kinase 1; ERBB ERBB1 HER1 PIG61 mENA avian erythroblastic leukemia viral (v-erb-b) oncogene homolog cell growth inhibiting protein 40 cell proliferation-inducing protein 61 proto-oncogene c-ErbB-1 receptor tyrosine-protein kinase erbB-1 epidermal growth f; Proto-oncogene c-ErbB-1; Receptor tyrosine-protein kinase erbB-1
Gene Aliases: EGFR; ERBB; ERBB1; HER1; mENA; NISBD2; PIG61
UniProt ID: (Human) P00533
Entrez Gene ID: (Human) 1956
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.