Invitrogen
Manufacturer Code:PA524584
Catalog # PIPA524584
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1004-1033 amino acids from the C-terminal region of human EGFR-S1026 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: avian erythroblastic leukemia viral (v-erb-b) oncogene homolog; cell growth inhibiting protein 40; cell proliferation-inducing protein 61; Epidermal growth factor receptor; erb-b2 receptor tyrosine kinase 1; ERBB ERBB1 HER1 PIG61 mENA avian erythroblastic leukemia viral (v-erb-b) oncogene homolog cell growth inhibiting protein 40 cell proliferation-inducing protein 61 proto-oncogene c-ErbB-1 receptor tyrosine-protein kinase erbB-1 epidermal growth f; Proto-oncogene c-ErbB-1; Receptor tyrosine-protein kinase erbB-1
Gene Aliases: EGFR; ERBB; ERBB1; HER1; mENA; NISBD2; PIG61
UniProt ID: (Human) P00533
Entrez Gene ID: (Human) 1956
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.