Invitrogen
Manufacturer Code:MA515606
Catalog # PIMA515606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4E11 |
Immunogen | Purified recombit fragment of human EGF expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: beta-urogastrone; EGF; Epidermal growth factor; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); URG HOMG4 EGF; Urogastrone
Gene Aliases: AI790464; EGF; HOMG4; URG
UniProt ID: (Human) P01133, (Mouse) Q6P9J2
Entrez Gene ID: (Human) 1950, (Mouse) 13645
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.