Invitrogen
Manufacturer Code:PA585447
Catalog # PIPA585447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human EFHA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110008L20Rik antibody FLJ25016 antibody FLJ34588 antibody EFHA1 antibody EF hand domain family A1 antibody EF-hand domain-containing family member A1 antibody EF hand domain family member A1 antibody Smhs2 homolog antibody EF-hand domain family member A1 antibody; Calcium uptake protein 2, mitochondrial; EF hand domain family A1; EF hand domain family, member A1; EF-hand domain family member A1; EF-hand domain family, member A1; EF-hand domain-containing family member A1; Smhs2 homolog
Gene Aliases: 09C01; 1110008L20Rik; 4833427E09Rik; AU041871; EFHA1; MICU2; RGD1309934; Smhs2
UniProt ID: (Human) Q8IYU8, (Rat) Q99P63, (Mouse) Q8CD10
Entrez Gene ID: (Human) 221154, (Rat) 171433, (Mouse) 68514
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.