Novus Biologicals
Manufacturer Code:NBP216278
Catalog # NBP216278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human EFHA1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110008L20Rik EF hand domain family A1 EF hand domain family member A1 EF-hand domain family member A1 EF-hand domain-containing family member A1 FLJ25016 FLJ34588 Smhs2 homolog; Calcium uptake protein 2, mitochondrial; EF hand domain family A1; EF hand domain family, member A1; EF-hand domain family member A1; EF-hand domain family, member A1; EF-hand domain-containing family member A1; Smhs2 homolog
Gene Aliases: 1110008L20Rik; EFHA1; MICU2
UniProt ID: (Human) Q8IYU8
Entrez Gene ID: (Human) 221154
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.