Abnova
Manufacturer Code:H00221154K
Catalog # 89-206-675
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human EFHA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human EFHA1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 1110008L20Rik FLJ25016 FLJ34588; Calcium uptake protein 2, mitochondrial; EF hand domain family A1; EF hand domain family, member A1; EF-hand domain family member A1; EF-hand domain family, member A1; EF-hand domain-containing family member A1; Smhs2 homolog
Gene Aliases: 1110008L20Rik; EFHA1; MICU2
UniProt ID: (Human) Q8IYU8
Entrez Gene ID: (Human) 221154
Molecular Function: calcium-binding protein calmodulin intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.