Invitrogen
Manufacturer Code:PA578555
Catalog # PIPA578555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human SELB. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EFSEC eukaryotic elongation factor selenocysteine-tRNA-specific EEFSEC; elongation factor for selenoprotein translation; Elongation factor sec; Eukaryotic elongation factor, selenocysteine-tRNA-specific; MJ0495-like protein SelB; mSelB; selenocysteine (Sec)-specific eukaryotic elongation factor; Selenocysteine-specific elongation factor
Gene Aliases: EEFSEC; EFSEC; sec; SELB
UniProt ID: (Human) P57772, (Mouse) Q9JHW4
Entrez Gene ID: (Human) 60678, (Mouse) 65967
Molecular Function:
G-protein
RNA binding protein
enzyme modulator
hydrolase
nucleic acid binding
translation elongation factor
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.