Novus Biologicals
Manufacturer Code:NBP2848370.1ML
Catalog # NB032765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human EEFSEC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: elongation factor for selenoprotein translation; Elongation factor sec; eukaryotic elongation factor selenocysteine-tRNA-specific; Eukaryotic elongation factor, selenocysteine-tRNA-specific; selenocysteine (Sec)-specific eukaryotic elongation factor; Selenocysteine-specific elongation factor
Gene Aliases: EEFSEC; EFSEC; SELB
UniProt ID: (Human) P57772
Entrez Gene ID: (Human) 60678
Molecular Function: G-protein RNA binding protein enzyme modulator hydrolase nucleic acid binding translation elongation factor translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.