Invitrogen
Manufacturer Code:PA5115163
Catalog # PIPA5115163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human eEF2K. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C86191 Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase calmodulin-dependent protein kinase III EC 2.7.11.20 eEF-2 kinase Eef2k eEF-2K EF2K elongation factor 2 kinase elongation factor-2 kinase euk eukaroytic elongation factor 2 kinase eukaryotic elongation factor 2 kinase eukaryotic elongation factor-2 kinase HSU93850 kinase eEF2K SMEF2K; Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase; calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase; calmodulin-dependent protein kinase III; eEF-2 kinase; eEF-2K; elongation factor 2 kinase; elongation factor-2 kinase; eukaroytic elongation factor 2 kinase; Eukaryotic elongation factor 2 kinase; eukaryotic elongation factor-2 kinase
Gene Aliases: C86191; eEF-2K; EEF2K; HSU93850; SMEF2K
UniProt ID: (Human) O00418, (Mouse) O08796, (Rat) P70531
Entrez Gene ID: (Human) 29904, (Mouse) 13631, (Rat) 25435
Molecular Function:
Hsp70 family chaperone
chaperone
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.