Novus Biologicals
Manufacturer Code:NB10087019
Catalog # NB10087019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 675 and 725 of human calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase (EEF2K ) using the numbering given in entry NP_037434.1 (GeneID 29904). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase; Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase EC 2.7.11 EC 2.7.11.20 eEF-2 kinase eEF-2Kcalmodulin-dependent protein kinase III elongation factor-2 kinase eukaroytic elongation factor 2 kinase eukaryotic elongation factor 2 kinase eukaryotic elongation factor-2 kinase HSU93850 MGC45041; calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase; calmodulin-dependent protein kinase III; eEF-2 kinase; elongation factor-2 kinase; eukaroytic elongation factor 2 kinase; Eukaryotic elongation factor 2 kinase; eukaryotic elongation factor-2 kinase
Gene Aliases: eEF-2K; EEF2K; HSU93850
UniProt ID: (Human) O00418
Entrez Gene ID: (Human) 29904
Molecular Function: Hsp70 family chaperone chaperone kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.