Invitrogen
Manufacturer Code:PA576656
Catalog # PIPA576656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human EEF1D. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY CO 4Antigen NY-CO-4EEF 1DEEF1DEF 1 deltaEF 1DEF-1-deltaEF1DEF1D&lowbarHUMANElongation factor 1 deltaElongation factor 1-deltaEukaryotic translation elongation factor 1 deltaGuanine nucleotide exchange protein; Antigen NY-CO-4; EF-1-delta; Elongation factor 1-delta; eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein); guanine nucleotide exchange protein; translation elongation factor 1-delta subunit
Gene Aliases: EEF1D; EF-1-delta; EF-1D; EF1D; FP1047
UniProt ID: (Human) P29692, (Mouse) P57776, (Rat) Q68FR9
Entrez Gene ID: (Human) 1936, (Mouse) 66656, (Rat) 300033
Molecular Function:
RNA binding protein
nucleic acid binding
translation elongation factor
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.