Novus Biologicals
Manufacturer Code:NBP191861
Catalog # NBP191861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SGTSGDHGELVVRIASLEVENQSLRGVVQELQQAISKLEARLNVLEKSSPGHRATAPQTQHVSPMRQVEPPAKKPATPAEDDED |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-4; Antigen NY-CO-4 EF-1D EF-1-delta EF1DFP1047 elongation factor 1-delta eukaryotic translation elongation factor 1 delta (guanine nucleotide exchangeprotein) FLJ20897 guanine nucleotide exchange protein; EF-1-delta; Elongation factor 1-delta; eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein); guanine nucleotide exchange protein
Gene Aliases: EEF1D; EF-1D; EF1D; FP1047
UniProt ID: (Human) P29692
Entrez Gene ID: (Human) 1936
Molecular Function:
RNA binding protein
nucleic acid binding
translation elongation factor
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.