Proteintech
Manufacturer Code:100952AP150UL
Catalog # 50-172-6019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EEF1B2 Fusion Protein Ag0135 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EEF1B EEF1B1 EEF1B2 EF 1 beta EF1B Elongation factor 1 beta; EF-1-beta; Elongation factor 1-beta; eukaryotic translation elongation factor 1 beta 1; eukaryotic translation elongation factor 1-beta homolog
Gene Aliases: 2810017J07Rik; EEF1B; EEF1B1; EEF1B2; EF1B
UniProt ID: (Human) P24534, (Mouse) O70251
Entrez Gene ID: (Human) 1933, (Rat) 363241, (Mouse) 55949
Molecular Function: RNA binding protein nucleic acid binding translation elongation factor translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.