Invitrogen
Manufacturer Code:MA535127
Catalog # PIMA535127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0626 |
Immunogen | A synthesized peptide derived from human eEF1A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CCS3 CCS-3 EE1A1 EEF-1 EEF1A Eef1a1 eEF1A-1 Eef1a2 Eef1a2l1 eef1as eef1a-s EF1A EF1A1 EF1alpha EF-1-alpha-1 EF-1-alpha-S EF-Tu Elongation factor 1-alpha 1 Elongation factor 1-alpha somatic form Elongation factor Tu Eukaryotic elongation factor 1 A-1 GRAF 1EF GRAF-1EF HNGC:16303 LENG7 Leukocyte receptor cluster member 7 PTI1 SI XELAEV_18029462mg; cervical cancer suppressor 3; CTCL tumor antigen; eEF1A-1; EF-1 alpha2; EF-1-alpha-1; EF-Tu; EF1a-like protein; elongation factor 1 A-1; elongation factor 1 alpha subunit; Elongation factor 1-alpha 1; Elongation factor Tu; Eukaryotic elongation factor 1 A-1; eukaryotic translation elongation factor 1 alpha 1-like 14; eukaryotic translation elongation factor 1 alpha 2 like 1; glucocorticoid receptor AF-1 specific elongation factor; leukocyte receptor cluster (LRC) member 7; Leukocyte receptor cluster member 7; prostate tumor-inducing protein 1; translation elongation factor 1 alpha 1-like 14
Gene Aliases: CCS-3; CCS3; EE1A1; EEF-1; EEF1A; eEF1A-1; EEF1A1; Eef1a2; Eef1a2l1; EF-Tu; EF1A; GRAF-1EF; HNGC:16303; LENG7; PTI1; SI
UniProt ID: (Human) P68104, (Mouse) P10126, (Rat) P62630
Entrez Gene ID: (Human) 1915, (Mouse) 13627, (Rat) 171361
Molecular Function: G-protein RNA binding protein enzyme modulator hydrolase nucleic acid binding translation elongation factor translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.