Invitrogen
Manufacturer Code:MA535927
Catalog # PIMA535927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT885 |
Immunogen | Recombinant protein encompassing a sequence within the center region of Human EED. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Eed embryonic ectoderm development Embryonic ectoderm development protein ENSMUSG00000039373 HEED l(7)5Rn l7Rn5 lethal Chr 7 Rinchik 5 lusk Polycomb protein EED WAIT1 WAIT-1 WD protein associating with integrin cytoplasmic tails 1; Embryonic ectoderm development protein; hEED; lethal, Chr 7, Rinchik 5; Polycomb protein EED; WAIT-1; WD protein associating with integrin cytoplasmic tails 1
Gene Aliases: EED; ENSMUSG00000039373; HEED; l(7)5Rn; l7Rn5; lusk; WAIT1
UniProt ID: (Human) O75530, (Mouse) Q921E6
Entrez Gene ID: (Human) 8726, (Mouse) 13626, (Rat) 293104
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.