Invitrogen
Manufacturer Code:PA592427
Catalog # PIPA592427
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-300 of human EED |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Embryonic ectoderm development protein; hEED; lethal, Chr 7, Rinchik 5; Polycomb protein EED; WAIT-1; WD protein associating with integrin cytoplasmic tails 1
Gene Aliases: EED; ENSMUSG00000039373; HEED; l(7)5Rn; l7Rn5; lusk; WAIT1
UniProt ID: (Human) O75530, (Mouse) Q921E6
Entrez Gene ID: (Human) 8726, (Mouse) 13626
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.