Novus Biologicals
Manufacturer Code:NBP281819
Catalog # NB017130
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EED antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human EED. The immunogen is located within amino acids 30 - 80 of EED. Amino Acid Squence: ESGTNTERPDTPTNTPNA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: embryonic ectoderm development HEED polycomb protein EED WAIT1 WAIT-1 WD protein associating with integrin cytoplasmic tails 1; Embryonic ectoderm development protein; hEED; Polycomb protein EED; WAIT-1; WD protein associating with integrin cytoplasmic tails 1
Gene Aliases: EED; HEED; WAIT1
UniProt ID: (Human) O75530
Entrez Gene ID: (Human) 8726
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.