R&D Systems
Manufacturer Code:AF5827
Catalog # AF5827
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human EED Ser2-Cys150 Accession # O75530 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: embryonic ectoderm development HEED polycomb protein EED WAIT1 WAIT-1 WD protein associating with integrin cytoplasmic tails 1; Embryonic ectoderm development protein; hEED; Polycomb protein EED; WAIT-1; WD protein associating with integrin cytoplasmic tails 1
Gene Aliases: EED; HEED; WAIT1
UniProt ID: (Human) O75530
Entrez Gene ID: (Human) 8726
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.