Proteintech
Manufacturer Code:168181AP150UL
Catalog # 50-173-1198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EED Fusion Protein Ag10358 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EED embryonic ectoderm development HEED Polycomb protein EED WAIT 1 WAIT1; Embryonic ectoderm development protein; hEED; Polycomb protein EED; WAIT-1; WD protein associating with integrin cytoplasmic tails 1
Gene Aliases: EED; HEED; WAIT1
UniProt ID: (Human) O75530
Entrez Gene ID: (Human) 8726
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.