R&D Systems
Manufacturer Code:FAB4496R100UG
Catalog # FAB4496R100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 671917 |
Immunogen | NS0 mouse myeloma cell line transfected with human EDNRB/Endothelin R Type B Accession &num P24530 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ABCDS EDNRB endothelin B receptor Endothelin receptor non-selective type endothelin receptor type B ETB ET-B ETBR ET-BR ETRB HSCR HSCR2 WS4A; Endothelin receptor non-selective type; endothelin receptor subtype B1; Endothelin receptor type B; ET-B
Gene Aliases: ABCDS; EDNRB; ET-B; ET-BR; ETB; ETB1; ETBR; ETRB; HSCR; HSCR2; WS4A
UniProt ID: (Human) P24530
Entrez Gene ID: (Human) 1910
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.