R&D Systems
Manufacturer Code:BAF4496
Catalog # BAF4496
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human EDNRB/Endothelin R Glu27-Lys101 Accession # P24530 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: ABCDS EDNRB endothelin B receptor Endothelin receptor non-selective type endothelin receptor type B ETB ET-B ETBR ET-BR ETRB HSCR HSCR2 WS4A; Endothelin receptor non-selective type; endothelin receptor subtype B1; Endothelin receptor type B; ET-B
Gene Aliases: ABCDS; EDNRB; ET-B; ET-BR; ETB; ETB1; ETBR; ETRB; HSCR; HSCR2; WS4A
UniProt ID: (Human) P24530
Entrez Gene ID: (Human) 1910
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.