Novus Biologicals
Manufacturer Code:NBP133614
Catalog # NBP133614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Endothelin A Receptor. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: endothelin receptor subtype A; Endothelin receptor type A; endothelin receptor type A endothelin-1 receptor endothelin-1-specific receptor ET1-specific type ETAR ETRA G protein-coupled receptor hET-AR; Endothelin-1 receptor; endothelin-1-specific receptor; ET-A; G protein-coupled receptor
Gene Aliases: EDNRA; ET-A; ETA; ETA-R; ETAR; ETRA; hET-AR; MFDA
UniProt ID: (Human) Q8TBH2
Entrez Gene ID: (Human) 1909
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.