Abnova
Manufacturer Code:PAB26205
Catalog # 89-127-844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 18 amino acid at extracellular domain of human EDNRA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of EDNRA.
For Research Use Only
Protein Aliases: endothelin receptor subtype A; Endothelin receptor type A; Endothelin-1 receptor; endothelin-1-specific receptor; ET-A; ETA; ETRA; G protein-coupled receptor
Gene Aliases: EDNRA; ET-A; ETA; ETA-R; ETAR; ETRA; hET-AR; MFDA
UniProt ID: (Human) Q8TBH2
Entrez Gene ID: (Human) 1909
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.