Abnova
Manufacturer Code:H00001908A01
Catalog # 89-104-415
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. to 238 a.a) full-length recombinant protein with GST tag.,EDN3 (AAH08876 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant EDN3.
For Research Use Only
Protein Aliases: Endothelin-3; ET-3; ET3; MGC15067; MGC61498; PPET3; Preproendothelin-3
Gene Aliases: EDN3; ET-3; ET3; HSCR4; PPET3; WS4B
UniProt ID: (Human) P14138
Entrez Gene ID: (Human) 1908
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.