Abnova
Manufacturer Code:MAB7737
Catalog # 89-113-188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 3G10 |
Immunogen | Recombinant protein (conjugated with BSA) corresponding to human EDN1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against partial recombinant EDN1.
For Research Use Only
Protein Aliases: Big endothelin-1; Endothelin-1; ET-1; ET1 HDLCQ7; PPET1; Preproendothelin-1
Gene Aliases: ARCND3; EDN1; ET1; HDLCQ7; PPET1; QME
UniProt ID: (Human) P05305
Entrez Gene ID: (Human) 1906
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.