Invitrogen
Manufacturer Code:PA577744
Catalog # PIPA577744
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)DYVGKLAGRLRD corresponding to amino acid residues 23-34 of mouse Sphingosine 1-phosphate receptor 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiac sphingosine-1-phosphate specific receptor; Endothelial differentiation G-protein coupled receptor 3; endothelial differentiation sphingolipid G-protein-coupled receptor 3; endothelial differentiation, sphingolipid G-protein-coupled receptor, 3; G protein-coupled receptor, endothelial differentiation gene-3; Lysophospholipid receptor B3; S1P receptor 3; S1P receptor Edg-3; S1P receptor EDG3; S1PR3 Edg3 Lysophospholipid receptor 3; Sphingosine 1-phosphate receptor 3; Sphingosine 1-phosphate receptor Edg-3
Gene Aliases: AI132464; EDG-3; EDG3; Edg3l; LPB3; S1P3; S1PR3
UniProt ID: (Human) Q99500, (Mouse) Q9Z0U9
Entrez Gene ID: (Human) 1903, (Mouse) 13610, (Rat) 306792
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.