Invitrogen
Manufacturer Code:PA523225
Catalog # PIPA523225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (326 SPIQPALDPSRSKSSSSNNSS 346) of Sphingolipid Receptor Edg3/S1P3 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EDG-3 EDG3 LPB3 S1P3 G protein-coupled receptor endothelial differentiation gene-3 S1P receptor 3 S1P receptor EDG3 S1P receptor Edg-3 endothelial differentiation G-protein coupled receptor 3 endothelial differentiation sphingolipid G-protein-coupled receptor 3 sphingosine 1-phosphate receptor 3 sphingosine 1-phosphate receptor Edg-3; Endothelial differentiation G-protein coupled receptor 3; endothelial differentiation sphingolipid G-protein-coupled receptor 3; endothelial differentiation, sphingolipid G-protein-coupled receptor, 3; G protein-coupled receptor, endothelial differentiation gene-3; Lysophospholipid receptor B3; S1P receptor 3; S1P receptor Edg-3; S1P receptor EDG3; Sphingosine 1-phosphate receptor 3; Sphingosine 1-phosphate receptor Edg-3
Gene Aliases: AI132464; EDG-3; EDG3; LPB3; S1P3; S1PR3
UniProt ID: (Human) Q99500, (Mouse) Q9Z0U9
Entrez Gene ID: (Human) 1903, (Mouse) 13610
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.