Proteintech
Manufacturer Code:204421AP150UL
Catalog # 50-173-2891
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EDG2 Fusion Protein Ag14253 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: edg 2 EDG2 Gpcr26 GPR26 LPA 1 LPA receptor 1 LPA1 LPAR1 Mrec1.3 rec.1.3 vzg 1 VZG1; endothelial differentiation lysophosphatidic acid G-protein-coupled receptor 2; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 2; G-protein coupled receptor 26; LPA receptor 1; LPA-1; Lysophosphatidic acid receptor 1; Lysophosphatidic acid receptor Edg-2; Rec1.3; ventricular zone gene 1; VZG-1
Gene Aliases: AI326300; edg-2; EDG2; Gpcr26; Gpcr91; GPR26; Kdt2; LPA1; LPAR1; Mrec1.3; rec.1.3; vzg-1; VZG1
UniProt ID: (Human) Q92633, (Rat) P61794, (Mouse) P61793
Entrez Gene ID: (Human) 1902, (Rat) 116744, (Mouse) 14745
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.