OriGene
Manufacturer Code:CF810789
Catalog # 50-167-0307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7D9 |
Immunogen | Full length human recombit protein of human EDF1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CFAP280 EDF-1 MBF1; EDF-1; Endothelial differentiation-related factor 1; MBF1; multiprotein bridging factor 1; Multiprotein-bridging factor 1
Gene Aliases: CFAP280; EDF-1; EDF1; MBF1
UniProt ID: (Human) O60869
Entrez Gene ID: (Human) 8721
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.