Invitrogen
Manufacturer Code:PA541467
Catalog # PIPA541467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human EDF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calmodulin-associated peptide 19; calmodulin-associated peptide-19; CAP-19; EDF-1; Endothelial differentiation-related factor 1; Endothelial differentiation-related factor 1 EDF-1 MBF1 MGC9058; endothelial differentiation-related factor 1 homolog; hypothetical protein 1-9; MBF1; multiprotein bridging factor 1; Multiprotein-bridging factor 1
Gene Aliases: 0610008L11Rik; AA409425; CAP-19; CFAP280; EDF-1; EDF1; MBF1
UniProt ID: (Bovine) Q3T0V7, (Human) O60869, (Rat) P69736, (Mouse) Q9JMG1
Entrez Gene ID: (Bovine) 515380, (Horse) 100068036, (Human) 8721, (Dog) 480674, (Guinea pig) 100733428, (Rat) 296570, (Mouse) 59022
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.