Invitrogen
Manufacturer Code:PA5116424
Catalog # PIPA5116424
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 122-146 amino acids from the C-terminal region of (DANRE)edf1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610008L11Rik AA409425 Calmodulin-associated peptide 19 calmodulin-associated peptide-19 CAP-19 CFAP280 Edf1 EDF-1 endothelial differentiation related factor 1 endothelial differentiation-related factor 1 endothelial differentiation-related factor 1 homolog hypothetical protein 1-9 MBF1 MGC9058 multiprotein bridging factor 1 multiprotein-bridging factor 1; EDF-1; Endothelial differentiation-related factor 1 homolog
Gene Aliases: edf1; zgc:73192; zgc:86829
UniProt ID: (Zebrafish) Q6PBY3
Entrez Gene ID: (Zebrafish) 793036
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.