Proteintech
Manufacturer Code:124191AP150UL
Catalog # 50-172-7968
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EDF1 Fusion Protein Ag3063 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EDF 1 EDF1 MBF1 Multiprotein bridging factor 1; EDF-1; Endothelial differentiation-related factor 1; hypothetical protein 1-9; MBF1; multiprotein bridging factor 1; Multiprotein-bridging factor 1
Gene Aliases: 0610008L11Rik; AA409425; CFAP280; EDF-1; EDF1; MBF1
UniProt ID: (Human) O60869, (Mouse) Q9JMG1
Entrez Gene ID: (Human) 8721, (Mouse) 59022
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.