Invitrogen
Manufacturer Code:PA5115159
Catalog # PIPA5115159
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human EDEM3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310050N11Rik Alpha-12-mannosidase EDEM3 C1orf22 EDEM3 ER degradation enhancer mannosidase alpha-like 3 ER degradation enhancing alpha-mannosidase like protein 3 ER degradation-enhancing alpha-mannosidase-like 3 ER degradation-enhancing alpha-mannosidase-like protein 3 ER degradation-enhancing -mannosidase-like protein 3 RGD1561496; Alpha-1,2-mannosidase EDEM3; ER degradation enhancer, mannosidase alpha-like 3; ER degradation-enhancing -mannosidase-like protein 3; ER degradation-enhancing alpha-mannosidase-like 3; ER degradation-enhancing alpha-mannosidase-like protein 3
Gene Aliases: 2310050N11Rik; C1orf22; EDEM3
UniProt ID: (Human) Q9BZQ6, (Mouse) Q2HXL6
Entrez Gene ID: (Human) 80267, (Mouse) 66967
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.