Novus Biologicals
Manufacturer Code:NBP258282
Catalog # NBP258282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SSQEVDLVDQESSEENSLNSHPESLSLADMDNAASISPSEQTSNPTENHETTNLNGECTDLDNQLQEQSETEEDSNPNVSWGKKVQPIDSILADWNEDIEAFEMMEKDE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,2-mannosidase EDEM3; C1orf22 chromosome 1 open reading frame 22 ER degradation enhancer mannosidase alpha-like 3 ER degradation-enhancing alpha-mannosidase-like 3 ER degradation-enhancing -mannosidase-like protein 3; ER degradation enhancer, mannosidase alpha-like 3; ER degradation-enhancing -mannosidase-like protein 3; ER degradation-enhancing alpha-mannosidase-like 3; ER degradation-enhancing alpha-mannosidase-like protein 3
Gene Aliases: C1orf22; EDEM3
UniProt ID: (Human) Q9BZQ6
Entrez Gene ID: (Human) 80267
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.