Proteintech
Manufacturer Code:112411AP150UL
Catalog # 50-172-6995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EDEM2 Fusion Protein Ag1755 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf31 C20orf49 EDEM2; ER degradation enhancer, mannosidase alpha-like 2; ER degradation-enhancing alpha-mannosidase-like 2; ER degradation-enhancing alpha-mannosidase-like protein 2; ER degradation-enhancing-mannosidase-like protein 2
Gene Aliases: 9530090G24Rik; AI327354; bA4204.1; C20orf31; C20orf49; EDEM2; RGD1303074; UNQ573/PRO1135
UniProt ID: (Human) Q9BV94, (Mouse) Q8BJT9
Entrez Gene ID: (Human) 55741, (Mouse) 108687, (Rat) 296304
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.